accessibility menu, dialog, popup

UserName

CH3OOCH3

CAS RN 931-94-2

IUPAC Name: 1,1-dimethoxycyclopentane
Synonyms: 1,1-dimethoxycyclopentane
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: AGWFDZMDKNQQHG-UHFFFAOYSA-N
SMILES: COC1(CCCC1)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results