accessibility menu, dialog, popup

UserName

CAS RN 930-27-8

IUPAC Name: 3-methylfuran
Synonyms: 3-methylfuran
Molecular Weight (g/mol): 82.1
Molecular Formula: C5H6O
InChi Key: KJRRQXYWFQKJIP-UHFFFAOYSA-N
SMILES: CC1=COC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)