accessibility menu, dialog, popup

UserName

CAS RN 93-62-9

NaNaOHNOOOO

CAS RN 93-62-9

IUPAC Name: disodium 2-[(carboxylatomethyl)(2-hydroxyethyl)amino]acetate
Synonyms: disodium hydroxyethyliminodiacetate
Molecular Weight (g/mol): 221.12
Molecular Formula: C6H9NNa2O5
InChi Key: ZTVCAEHRNBOTLI-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OCCN(CC([O-])=O)CC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results