accessibility menu, dialog, popup

UserName

CAS RN 93-29-8

CH3OCH3OCH3O(E)

CAS RN 93-29-8

IUPAC Name: 2-methoxy-4-[(1E)-prop-1-en-1-yl]phenyl acetate
Synonyms: isoeugenyl acetate
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: IUSBVFZKQJGVEP-SNAWJCMRSA-N
SMILES: COC1=CC(\C=C\C)=CC=C1OC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results