accessibility menu, dialog, popup

UserName

H2NSOHOO

CAS RN 93-00-5

IUPAC Name: 6-aminonaphthalene-2-sulfonic acid
Synonyms: bronner acid bronner's acid phenamine fast scarlet 4bgp broenner-saeure broenner acid broenner's acid pontamine fast scarlet 4BA 1,6-clev acid 1,6-clev's acid
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: SEMRCUIXRUXGJX-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=C(C=CC2=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results