CAS RN 929-75-9
CAS RN 929-75-9
IUPAC Name:
2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethan-1-amine
Synonyms:
2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethanamine
Molecular Weight (g/mol):
192.26
Molecular Formula:
C8H20N2O3
InChi Key:
NIQFAJBKEHPUAM-UHFFFAOYSA-N
SMILES:
NCCOCCOCCOCCN
1
–
1
of
1
results
1
1,11-Diamino-3,6,9-trioxaundecane 97.0+%, TCI America™
CAS: 929-75-9 Molecular Formula: C8H20N2O3 Molecular Weight (g/mol): 192.26 MDL Number: MFCD00628164 InChI Key: NIQFAJBKEHPUAM-UHFFFAOYSA-N Synonym: Bis[2-(2-aminoethoxy)ethyl] Ether, 3,6,9-Trioxaundecane-1,11-diamine PubChem CID: 70250 IUPAC Name: 2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethan-1-amine SMILES: NCCOCCOCCOCCN