accessibility menu, dialog, popup

UserName

CAS RN 928664-98-6

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
Molecular Weight (g/mol): 195.03
Molecular Formula: C9H14BNO3
InChi Key: LXCICYRNWIGDQA-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CON=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results