accessibility menu, dialog, popup

UserName

H3CCH3CH3OONHOHOOO

CAS RN 92828-64-3

IUPAC Name: 4-(benzyloxy)-3-{[(tert-butoxy)carbonyl]amino}-4-oxobutanoic acid
Synonyms: 4-(benzyloxy)-3-[(tert-butoxycarbonyl)amino]-4-oxobutanoic acid
Molecular Weight (g/mol): 323.35
Molecular Formula: C16H21NO6
InChi Key: LDRWTKQWSXGSTM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(O)=O)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)