CAS RN 927391-09-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methoxy-4-(tri-n-butylstannyl)thiazole, 96%, Thermo Scientific™
CAS: 927391-09-1 Molecular Formula: C16H31NOSSn Molecular Weight (g/mol): 404.2 MDL Number: MFCD07787372 InChI Key: JCZRGWAGRPABLT-UHFFFAOYSA-N Synonym: 2-methoxy-4-tributylstannyl thiazole,2-methoxy-4-tributylstannyl-1,3-thiazole,4-tributylstannyl-2-methoxythiazole,acmc-209rha,tributyl-2-methoxy-1,3-thiazol-4-yl stannane,2-methoxy-4-tri-n-butylstannyl thiazole,4-tri-n-butylstannyl-2-methoxy-1,3-thiazole PubChem CID: 2763264 IUPAC Name: tributyl-(2-methoxy-1,3-thiazol-4-yl)stannane SMILES: CCCC[Sn](CCCC)(CCCC)C1=CSC(=N1)OC