CAS RN 927-73-1
IUPAC Name:
4-chlorobut-1-ene
Synonyms:
4-chloro-1-butene
Molecular Weight (g/mol):
90.55
Molecular Formula:
C4H7Cl
InChi Key:
WKEVRZCQFQDCIR-UHFFFAOYSA-N
SMILES:
ClCCC=C
1
–
2
of
2
results
1
Promotions Available
4-Chloro-1-butene, 98%, Thermo Scientific Chemicals
4-Chloro-1-butene, 98%, Thermo Scientific Chemicals
CAS: 927-73-1 Molecular Formula: C4H7Cl Molecular Weight (g/mol): 90.55 InChI Key: WKEVRZCQFQDCIR-UHFFFAOYSA-N Synonym: 4-chloro-1-butene,1-butene, 4-chloro,1-but-3-enyl chloride,ksc487m9b PubChem CID: 13565 IUPAC Name: 4-chlorobut-1-ene SMILES: C=CCCCl
2
Promotions Available
4-Chloro-1-butene, 98%, Thermo Scientific™