CAS RN 926023-82-7
IUPAC Name:
N-(3-aminopropyl)-2-[(3-methylphenyl)methoxy]-N-[(thiophen-2-yl)methyl]benzamide hydrochloride
Synonyms:
N-(3-aminopropyl)-2-[(3-methylphenyl)methoxy]-N-(thiophen-2-ylmethyl)benzamide hydrochloride
Molecular Weight (g/mol):
430.99
Molecular Formula:
C23H27ClN2O2S
InChi Key:
UDXGBANGPYONOK-UHFFFAOYSA-N
SMILES:
Cl.CC1=CC(COC2=CC=CC=C2C(=O)N(CCCN)CC2=CC=CS2)=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ AMTB hydrochloride
CAS: 926023-82-7 Molecular Formula: C23H27ClN2O2S