accessibility menu, dialog, popup

UserName

CAS RN 92390-26-6

IUPAC Name: Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
Synonyms:
Molecular Weight (g/mol): 379.93
Molecular Formula: C18H27ClRu
InChi Key:
SMILES: Cl[Ru++].C1C\C=C/CC\C=C/1.Cc1c(C)c(C)c(C)c1C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results