CAS RN 923-61-5
CAS RN 923-61-5
IUPAC Name:
(2-aminoethoxy)[(2R)-2,3-bis(hexadecanoyloxy)propoxy]phosphinic acid
Synonyms:
DPPE dipalmitoyl phosphatidylethanolamine
Molecular Weight (g/mol):
691.97
Molecular Formula:
C37H74NO8P
InChi Key:
SLKDGVPOSSLUAI-PGUFJCEWSA-N
SMILES:
CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)OCCN)OC(=O)CCCCCCCCCCCCCCC
1
–
1
of
1
results
1
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine 96.0+%, TCI America™
CAS: 923-61-5 Molecular Formula: C37H74NO8P Molecular Weight (g/mol): 691.972 MDL Number: MFCD00036776 InChI Key: SLKDGVPOSSLUAI-PGUFJCEWSA-N Synonym: 1,2-Dihexadecanoyl-sn-glycero-3-phosphoethanolamine, Dipalmitoylphosphatidylethanolamine, 2-Aminoethyl (R)-2,3-Bis(hexadecanoyloxy)propyl Hydrogen Phosphate, L-DPPE PubChem CID: 445468 ChEBI: CHEBI:73127 IUPAC Name: [(2R)-3-[2-aminoethoxy(hydroxy)phosphoryl]oxy-2-hexadecanoyloxypropyl] hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)(O)OCCN)OC(=O)CCCCCCCCCCCCCCC