CAS RN 92243-74-8
IUPAC Name:
[4-(2-ethoxy-2-oxoethyl)phenyl]boronic acid
Synonyms:
4-(2-ethoxy-2-oxoethyl)phenylboronic acid
Molecular Weight (g/mol):
208.02
Molecular Formula:
C10H13BO4
InChi Key:
RDIRUZGSOKLANG-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC1=CC=C(C=C1)B(O)O
1
–
1
of
1
results
1
Promotions Available
4-[(Ethoxycarbonyl)methyl]phenylboronic acid, 97%
4-[(Ethoxycarbonyl)methyl]phenylboronic acid, 97%
CAS: 92243-74-8 Molecular Formula: C10H13BO4 Molecular Weight (g/mol): 208.02 MDL Number: MFCD08704683 InChI Key: RDIRUZGSOKLANG-UHFFFAOYSA-N IUPAC Name: [4-(2-ethoxy-2-oxoethyl)phenyl]boronic acid SMILES: CCOC(=O)CC1=CC=C(C=C1)B(O)O