CAS RN 91990-88-4
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 4-benzoylbenzoate
Synonyms:
2,5-dioxopyrrolidin-1-yl 4-benzoylbenzoate
Molecular Weight (g/mol):
323.3
Molecular Formula:
C18H13NO5
InChi Key:
MVQNJLJLEGZFGP-UHFFFAOYSA-N
SMILES:
O=C(ON1C(=O)CCC1=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
N-Succinimidyl 4-Benzoylbenzoate 98.0+%, TCI America™
CAS: 91990-88-4 Molecular Formula: C18H13NO5 Molecular Weight (g/mol): 323.30 MDL Number: MFCD00058572 InChI Key: MVQNJLJLEGZFGP-UHFFFAOYSA-N Synonym: 4-Benzoylbenzoic Acid N-Succinimidyl Ester PubChem CID: 2762595 IUPAC Name: 2,5-dioxopyrrolidin-1-yl 4-benzoylbenzoate SMILES: O=C(ON1C(=O)CCC1=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1