CAS RN 916791-97-4
IUPAC Name:
ethyl 5-tert-butyl-1H-pyrazole-3-carboxylate
Synonyms:
ethyl 5-tert-butyl-1H-pyrazole-3-carboxylate
Molecular Weight (g/mol):
196.25
Molecular Formula:
C10H16N2O2
InChi Key:
RCXMILPYEKVQLB-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=NNC(=C1)C(C)(C)C
1
–
1
of
1
results
1
Promotions Available
Ethyl 3-tert-butyl-1H-pyrazole-5-carboxylate, 97%
Ethyl 3-tert-butyl-1H-pyrazole-5-carboxylate, 97%
CAS: 916791-97-4 Molecular Formula: C10H16N2O2 Molecular Weight (g/mol): 196.25 MDL Number: MFCD00173782 InChI Key: RCXMILPYEKVQLB-UHFFFAOYSA-N Synonym: ethyl 3-tert-butyl-1h-pyrazole-5-carboxylate,5-tert-butyl-2h-pyrazole-3-carboxylic acid ethyl ester,5-tert-butyl-2h-pyrazole-3-carboxylic acid ethylester,ethyl 5-tert-butyl-1h-pyrazole-3-carboxylate,ethyl 5-tert-butyl-2h-pyrazole-3-carboxylate,1h-pyrazole-3-carboxylic acid, 5-1,1-dimethylethyl-, ethyl ester,maybridge3_006722,acmc-1cr14 PubChem CID: 2743509 IUPAC Name: ethyl 5-tert-butyl-1H-pyrazole-3-carboxylate SMILES: CCOC(=O)C1=NNC(=C1)C(C)(C)C