accessibility menu, dialog, popup

UserName

BNSOOH3CHOOH

CAS RN 913836-01-8

IUPAC Name: (5-methanesulfonylpyridin-3-yl)boronic acid
Synonyms: 5-methanesulfonylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 201
Molecular Formula: C6H8BNO4S
InChi Key: YGXNPQDXWDCWET-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CN=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results