accessibility menu, dialog, popup

UserName

CH3NNOOBr

CAS RN 911434-05-4

IUPAC Name: 5-bromo-2-methyl-3-nitropyridine
Synonyms: 5-bromo-2-methyl-3-nitropyridine
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: FZZLWWNOYMHSIS-UHFFFAOYSA-N
SMILES: CC1=NC=C(Br)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results