accessibility menu, dialog, popup

UserName

H2NNH2HN(S)NHOOOO

CAS RN 91000-69-0

IUPAC Name: (2S)-5-{[amino(iminiumyl)methyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoate
Synonyms: (2S)-5-{[amino(iminio)methyl]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoate
Molecular Weight (g/mol): 396.45
Molecular Formula: C21H24N4O4
InChi Key: DVBUCBXGDWWXNY-SFHVURJKSA-N
SMILES: NC(=[NH2+])NCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)