CAS RN 9052-95-3
CAS RN 9052-95-3
IUPAC Name:
1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
Synonyms:
2-ethylstyrene; divinylbenzene; styrene
Molecular Weight (g/mol):
366.55
Molecular Formula:
C28H30
InChi Key:
NWUYHJFMYQTDRP-UHFFFAOYSA-N
SMILES:
C=CC1=CC=CC=C1.CCC1=CC=CC=C1C=C.C=CC1=CC=CC=C1C=C
1
–
2
of
2
results
1
MilliporeSigma™ Supelco™ Diaion™ HP-20
Swelling in toluene is 30%
2
MilliporeSigma™ Supelco™ Diaion™ HP-20
Swelling in toluene is 30%