accessibility menu, dialog, popup

UserName

CAS RN 904696-60-2

IUPAC Name: lithium(1+) 2-[(morpholin-4-yl)methyl]benzoate
Synonyms: lithium(1+) 2-(morpholin-4-ylmethyl)benzoate
Molecular Weight (g/mol): 227.19
Molecular Formula: C12H14LiNO3
InChi Key: PKLIDBHLNQBFTO-UHFFFAOYSA-M
SMILES: [Li+].[O-]C(=O)C1=CC=CC=C1CN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)