accessibility menu, dialog, popup

UserName

CAS RN 903876-45-9

IUPAC Name: 3-{5,5,6-trimethylbicyclo[2.2.1]heptan-2-yl}cyclohexyl prop-2-enoate
Synonyms: 3-{5,5,6-trimethylbicyclo[2.2.1]heptan-2-yl}cyclohexyl prop-2-enoate
Molecular Weight (g/mol): 290.45
Molecular Formula: C19H30O2
InChi Key: CDBRNGRSVNBVLJ-UHFFFAOYNA-N
SMILES: CC1C2CC(CC2C2CCCC(C2)OC(=O)C=C)C1(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results