accessibility menu, dialog, popup

UserName

CAS RN 902836-82-2

IUPAC Name: 2-(4-chlorophenoxy)-6-fluorobenzaldehyde
Synonyms: 2-(4-chlorophenoxy)-6-fluorobenzaldehyde
Molecular Weight (g/mol): 250.65
Molecular Formula: C13H8ClFO2
InChi Key: JEPXYNGAXLVUMW-UHFFFAOYSA-N
SMILES: FC1=CC=CC(OC2=CC=C(Cl)C=C2)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results