accessibility menu, dialog, popup

UserName

CAS RN 90213-66-4

HNNNClCl

CAS RN 90213-66-4

IUPAC Name: 2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine
Synonyms: 2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine
Molecular Weight (g/mol): 188.01
Molecular Formula: C6H3Cl2N3
InChi Key: GHXBPCSSQOKKGB-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C2C=CNC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results