CAS RN 9013-20-1
CAS RN 9013-20-1
IUPAC Name:
1-{[(carboxymethyl)sulfanyl](3-chlorophenyl)methylidene}-1λ⁵-piperidin-1-ylium bromide
Synonyms:
1-{[(carboxymethyl)sulfanyl](3-chlorophenyl)methylidene}-1λ⁵-piperidin-1-ylium bromide
Molecular Weight (g/mol):
378.71
Molecular Formula:
C14H17BrClNO2S
InChi Key:
RTWACOLFHOBGCE-UHFFFAOYSA-N
SMILES:
[Br-].OC(=O)CSC(C1=CC(Cl)=CC=C1)=[N+]1CCCCC1
1
–
1
of
1
results
1
Streptavidin, Streptomyces avidinii, Thermo Scientific Chemicals
CAS: 9013-20-1 Molecular Formula: C14H17BrClNO2S Molecular Weight (g/mol): 378.709 MDL Number: MFCD00082035 InChI Key: RTWACOLFHOBGCE-UHFFFAOYSA-N PubChem CID: 51062757 IUPAC Name: 2-[(3-chlorophenyl)-piperidin-1-ium-1-ylidenemethyl]sulfanylacetic acid;bromide SMILES: C1CC[N+](=C(C2=CC(=CC=C2)Cl)SCC(=O)O)CC1.[Br-]