CAS RN 9012-72-0
CAS RN 9012-72-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ β-Glucan, Saccharomyces cerevisiae
Shown to induce oxidative burst response from NR8383 cells
2
Glucan from Black Yeast, TCI America™
CAS: 9012-72-0 MDL Number: MFCD00466924