missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ β-Glucan, Saccharomyces cerevisiae

Catalog No. 34621025MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34621025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34621025MG Supplier MilliporeSigma™ Supplier No. 34621025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Shown to induce oxidative burst response from NR8383 cells

  • A major structural element of the yeast cell wall having a β1,3-glucan linear structure with a small number of β1,6-glucan branches
  • Has been shown to induce an oxidative burst response from NR8383 cells
  • M.W. 200,000 (avg.). Insoluble in H2O. Forms suspension in basic solution

Specifications

CAS 9012-72-0
Color Off-white
Content And Storage Store at +15°C to +30°C
Purity ≥95% by beta-glucan
Quantity 25 mg
Form Solid