accessibility menu, dialog, popup

UserName

H3COOHCH3OOH3C

CAS RN 90-24-4

IUPAC Name: 1-(2-hydroxy-4,6-dimethoxyphenyl)ethan-1-one
Synonyms: xanthoxylin brevifolin (zanthoxylum) phloracetophenone dimethyl ether
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: FBUBVLUPUDBFME-UHFFFAOYSA-N
SMILES: COC1=CC(O)=C(C(C)=O)C(OC)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)