accessibility menu, dialog, popup

UserName

CAS RN 89980-69-8

IUPAC Name: bromo(3,4-dimethoxyphenyl)magnesium
Synonyms: bromo(3,4-dimethoxyphenyl)magnesium
Molecular Weight (g/mol): 241.37
Molecular Formula: C8H9BrMgO2
InChi Key: BHTPZWCAIHFRSY-UHFFFAOYSA-M
SMILES: COC1=CC=C([Mg]Br)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results