accessibility menu, dialog, popup

UserName

CAS RN 89972-77-0

IUPAC Name: 4-(4-methylphenyl)-6-(pyridin-2-yl)-2,2'-bipyridine
Synonyms: 4-(4-methylphenyl)-6-(pyridin-2-yl)-2,2'-bipyridine
Molecular Weight (g/mol): 323.4
Molecular Formula: C22H17N3
InChi Key: IDJYBUVHZHLIIT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results