accessibility menu, dialog, popup

UserName

CAS RN 89823-23-4

IUPAC Name: 6-({4'-cyano-[1,1'-biphenyl]-4-yl}oxy)hexyl prop-2-enoate
Synonyms: 6-({4'-cyano-[1,1'-biphenyl]-4-yl}oxy)hexyl prop-2-enoate
Molecular Weight (g/mol): 349.43
Molecular Formula: C22H23NO3
InChi Key: IGHSOWSFSFGPAZ-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results