accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOOOH

CAS RN 89813-47-8

IUPAC Name: 2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate
Synonyms: 2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 321.37
Molecular Formula: C17H23NO5
InChi Key: BEIPCYKSYYZEJH-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(O)CC1C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)