accessibility menu, dialog, popup

UserName

CAS RN 89794-02-5

CH3BrCl

CAS RN 89794-02-5

IUPAC Name: 4-bromo-2-chloro-1-methylbenzene
Synonyms: 4-bromo-2-chloro-1-methylbenzene
Molecular Weight (g/mol): 205.48
Molecular Formula: C7H6BrCl
InChi Key: LIFMTDJMLRECMX-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=C(Br)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results