accessibility menu, dialog, popup

UserName

BHOOH

CAS RN 89490-05-1

IUPAC Name: (cyclohex-1-en-1-yl)boronic acid
Synonyms: cyclohex-1-en-1-ylboronic acid
Molecular Weight (g/mol): 125.96
Molecular Formula: C6H11BO2
InChi Key: XZWQKJXJNKYMAP-UHFFFAOYSA-N
SMILES: OB(O)C1=CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results