CAS RN 892502-16-8
CAS RN 892502-16-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-bromo-6-(tetrahydropyran-4-yloxy)pyridine, 97%, Thermo Scientific™
CAS: 892502-16-8 Molecular Formula: C10H12BrNO2 Molecular Weight (g/mol): 258.115 MDL Number: MFCD12198112 InChI Key: VLEKIKTZHYMZEW-UHFFFAOYSA-N Synonym: 2-bromo-6-tetrahydropyran-4-yloxy pyridine,2-bromo-6-oxan-4-yloxy pyridine,2-bromo-6-oxan-4-yl oxy pyridine,2-2h-3,4,5,6-tetrahydropyran-4-yloxy-6-bromopyridine PubChem CID: 43811040 IUPAC Name: 2-bromo-6-(oxan-4-yloxy)pyridine SMILES: C1COCCC1OC2=NC(=CC=C2)Br