accessibility menu, dialog, popup

UserName

CH3HNON

CAS RN 892501-90-5

IUPAC Name: methyl({[4-(pyridin-2-yloxy)phenyl]methyl})amine
Synonyms: methyl({[4-(pyridin-2-yloxy)phenyl]methyl})amine
Molecular Weight (g/mol): 214.27
Molecular Formula: C13H14N2O
InChi Key: AFHNQZMRTDPSDR-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(OC2=CC=CC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)