CAS RN 89171-23-3
IUPAC Name:
potassium tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Synonyms:
potassium tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Molecular Weight (g/mol):
718.14
Molecular Formula:
C24BF20K
InChi Key:
GYBHRIJOPWTIKA-UHFFFAOYSA-N
SMILES:
[K+].FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
1
–
1
of
1
results
1
Promotions Available
Potassium tetrakis(pentafluorophenyl)borate, 97%
Potassium tetrakis(pentafluorophenyl)borate, 97%
CAS: 89171-23-3 Molecular Formula: C24BF20K Molecular Weight (g/mol): 718.14 MDL Number: MFCD06797410 InChI Key: GYBHRIJOPWTIKA-UHFFFAOYSA-N Synonym: potassium tetrakis pentafluorophenyl borate,potassium tetrakis pentafluorophe-nyl borate,tetrakis-pentafluorophenyl-potassium borate,potassium tetrakis 2,3,4,5,6-pentafluorophenyl boranuide PubChem CID: 23693577 IUPAC Name: potassium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide SMILES: [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.[K+]