accessibility menu, dialog, popup

UserName

CAS RN 891180-59-9

IUPAC Name: 2-bromo-3-fluorobenzaldehyde
Synonyms: 2-bromo-3-fluorobenzaldehyde
Molecular Weight (g/mol): 203.01
Molecular Formula: C7H4BrFO
InChi Key: CYJBMGYWRHGZBR-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C=O)=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results