accessibility menu, dialog, popup

UserName

NNNBrBrBr

CAS RN 890704-02-6

IUPAC Name: 4,7-dibromo-2-(6-bromohexyl)-2H-1,2,3-benzotriazole
Synonyms: 4,7-dibromo-2-(6-bromohexyl)-1,2,3-benzotriazole
Molecular Weight (g/mol): 439.98
Molecular Formula: C12H14Br3N3
InChi Key: VWPRIVLLHDPAJM-UHFFFAOYSA-N
SMILES: BrCCCCCCN1N=C2C(=N1)C(Br)=CC=C2Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results