accessibility menu, dialog, popup

UserName

CAS RN 88581-00-4

IUPAC Name: 4-(4-butylcyclohexyl)phenol
Synonyms: 4-(4-butylcyclohexyl)phenol
Molecular Weight (g/mol): 232.37
Molecular Formula: C16H24O
InChi Key: SJXVTMAQPHVBEG-UHFFFAOYSA-N
SMILES: CCCCC1CCC(CC1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results