CAS RN 885465-97-4
CAS RN 885465-97-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(1,3-Thiazol-2-yl)benzaldehyde, 97%, Thermo Scientific™
CAS: 885465-97-4 Molecular Formula: C10H7NOS Molecular Weight (g/mol): 189.232 MDL Number: MFCD06797781 InChI Key: YJMGIEKCDQXKKZ-UHFFFAOYSA-N Synonym: 3-thiazol-2-yl-benzaldehyde,3-1,3-thiazol-2-yl benzaldehyde,3-thiazol-2-yl benzaldehyde,benzaldehyde,3-2-thiazolyl,benzaldehyde, 3-2-thiazolyl PubChem CID: 18525725 IUPAC Name: 3-(1,3-thiazol-2-yl)benzaldehyde SMILES: C1=CC(=CC(=C1)C=O)C2=NC=CS2