accessibility menu, dialog, popup

UserName

SOONHNH2ClHCl

CAS RN 88519-57-7

IUPAC Name: N-(4-aminobutyl)-5-chloronaphthalene-2-sulfonamide hydrochloride
Synonyms: N-(4-aminobutyl)-5-chloronaphthalene-2-sulfonamide hydrochloride
Molecular Weight (g/mol): 349.27
Molecular Formula: C14H18Cl2N2O2S
InChi Key: QKAALLVQBOLELJ-UHFFFAOYSA-N
SMILES: Cl.NCCCCNS(=O)(=O)C1=CC2=CC=CC(Cl)=C2C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results