CAS RN 884507-14-6
CAS RN 884507-14-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Methyl-N-[(6-pyrrolidin-1-ylpyridin-2-yl)methyl]amine, 97%, Thermo Scientific™
CAS: 884507-14-6 Molecular Formula: C11H17N3 Molecular Weight (g/mol): 191.278 MDL Number: MFCD09025832 InChI Key: GGQYFTXRIMPZQO-UHFFFAOYSA-N Synonym: n-methyl-n-6-pyrrolidin-1-ylpyridin-2-yl methyl amine,2-pyridinemethanamine,n-methyl-6-1-pyrrolidinyl,n-methyl 6-pyrrolidin-1-yl pyridin-2-yl methanamine,methyl 6-pyrrolidin-1-yl pyridin-2-yl methyl amine,methyl 6-pyrrolidinyl 2-pyridyl methyl amine,2-pyridinemethanamine, n-methyl-6-1-pyrrolidinyl,n-methyl-1-6-1-pyrrolidinyl-2-pyridinyl methanamine,n-methyl-1-6-pyrrolidin-1-yl pyridin-2-yl methanamine PubChem CID: 24229485 IUPAC Name: N-methyl-1-(6-pyrrolidin-1-ylpyridin-2-yl)methanamine SMILES: CNCC1=NC(=CC=C1)N2CCCC2