CAS RN 882-06-4
IUPAC Name:
(2E)-3-(4-nitrophenyl)prop-2-enoic acid
Synonyms:
trans-P-nitrocinnamic acid
Molecular Weight (g/mol):
193.16
Molecular Formula:
C9H7NO4
InChi Key:
XMMRNCHTDONGRJ-ZZXKWVIFSA-N
SMILES:
OC(=O)\C=C\C1=CC=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
trans-4-Nitrocinnamic acid, 98+%
trans-4-Nitrocinnamic acid, 98+%
CAS: 882-06-4 Molecular Formula: C9H7NO4 Molecular Weight (g/mol): 193.158 MDL Number: MFCD00007381 InChI Key: XMMRNCHTDONGRJ-ZZXKWVIFSA-N Synonym: 4-nitrocinnamic acid,3-4-nitrophenyl acrylic acid,p-nitrocinnamic acid,cinnamic acid, p-nitro,2-propenoic acid, 3-4-nitrophenyl,e-3-4-nitrophenyl acrylic acid,3-4-nitrophenyl propenoic acid,trans-4-nitrocinnamic acid,ccris 1665,2e-3-4-nitrophenyl prop-2-enoic acid PubChem CID: 737157 SMILES: C1=CC(=CC=C1C=CC(=O)O)[N+](=O)[O-]