accessibility menu, dialog, popup

UserName

CAS RN 88038-94-2

CH3OO

CAS RN 88038-94-2

IUPAC Name: 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate
Synonyms: 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate
Molecular Weight (g/mol): 251.39
Molecular Formula: C16H27O2
InChi Key: JXPGQFKJNKWDKP-UHFFFAOYSA-M
SMILES: CCCC1CCC(CC1)C1CCC(CC1)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results