accessibility menu, dialog, popup

UserName

OHNOONOONOO

CAS RN 88-89-1

IUPAC Name: 2,4,6-trinitrophenol
Synonyms: 2,4,6-trinitrophenol acide picrique
Molecular Weight (g/mol): 229.1
Molecular Formula: C6H3N3O7
InChi Key: OXNIZHLAWKMVMX-UHFFFAOYSA-N
SMILES: OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
Grade 
  • (3)
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (6)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)