missing translation for 'onlineSavingsMsg'
Learn More

StatLab MasterTech Saturated Picric Acid Solution

Catalog No. 23640006
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23640006 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23640006 Supplier StatLab Supplier No. AHSPA100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

Used as a decalcifying agent for small tissue samples and as a component in fixatives and staining applications.

  • Used in histology laboratories as a component of fixatives, a decalcifying agent for small tissue samples, and an essential ingredient in selected differential staining techniques.
  • Exhibits strong protein-precipitating properties, an acidic nature, and the ability to function as an acidic dye, making it valuable for effective preparation, fixation, and analysis of biological tissues.
  • Supplied as a MasterTech Special Stain Kit component, ensuring compatibility, consistency, and reliable performance in professional histology and pathology workflows.

Specifications

Physical Form Solution
CAS 88-89-1
Color Yellow
Quantity 100 mL
Grade Laboratory