CAS RN 88-27-7
IUPAC Name:
2,6-di-tert-butyl-4-[(dimethylamino)methyl]phenol
Synonyms:
ethyl 703
Molecular Weight (g/mol):
263.43
Molecular Formula:
C17H29NO
InChi Key:
VMZVBRIIHDRYGK-UHFFFAOYSA-N
SMILES:
CN(C)CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
1
–
1
of
1
results
1
2,6-Di-tert-butyl-4-dimethylaminomethylphenol 98.0+%, TCI America™
CAS: 88-27-7 Molecular Formula: C17H29NO Molecular Weight (g/mol): 263.425 MDL Number: MFCD00026283 InChI Key: VMZVBRIIHDRYGK-UHFFFAOYSA-N Synonym: ethyl 703,2,6-di-tert-butyl-4-dimethylaminomethyl phenol,n,n-dimethyl-3,5-di-tert-butyl-4-hydroxybenzylamine,agidol 3,f 1 antioxidant,ethyl antioxidant 703,2,6-di-tert-butyl-4-dimethylamino methylphenol,unii-8tk6vr2kyx,2,6-di-tert-butyl-alpha-dimethylamino-p-cresol,n-3,5-di-tert-butyl-4-hydroxybenzyl dimethylamine PubChem CID: 66609 IUPAC Name: 2,6-ditert-butyl-4-[(dimethylamino)methyl]phenol SMILES: CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CN(C)C