accessibility menu, dialog, popup

UserName

CAS RN 88-24-4

IUPAC Name: 2-tert-butyl-6-[(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methyl]-4-ethylphenol
Synonyms: 2-tert-butyl-6-[(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methyl]-4-ethylphenol
Molecular Weight (g/mol): 368.56
Molecular Formula: C25H36O2
InChi Key: GPNYZBKIGXGYNU-UHFFFAOYSA-N
SMILES: CCC1=CC(=C(O)C(CC2=C(O)C(=CC(CC)=C2)C(C)(C)C)=C1)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results