accessibility menu, dialog, popup

UserName

CH3NHOCH3H3CNNHNNOCH3

CAS RN 878739-06-1

IUPAC Name: 3-(1-cyano-1-methylethyl)-N-{4-methyl-3-[(3-methyl-4-oxo-3,4-dihydroquinazolin-6-yl)amino]phenyl}benzamide
Synonyms: 3-(1-cyano-1-methylethyl)-N-{4-methyl-3-[(3-methyl-4-oxoquinazolin-6-yl)amino]phenyl}benzamide
Molecular Weight (g/mol): 451.53
Molecular Formula: C27H25N5O2
InChi Key: ZGBGPEDJXCYQPH-UHFFFAOYSA-N
SMILES: CN1C=NC2=CC=C(NC3=C(C)C=CC(NC(=O)C4=CC(=CC=C4)C(C)(C)C#N)=C3)C=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results